Learn More
Biomatik Corporation RNF126 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07882
Encompass_Preferred
Rnf126 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to RNF126. The Rnf126 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human E3 Ubiquitin-Protein Ligase Rnf126 Protein (39-225Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: RNF126. Accession Number: Q9BV68. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7467