Learn More
Biomatik Corporation USP6 anti-Human, Rat, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07898
Encompass_Preferred
Usp6 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to USP6. The Usp6 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Ubiquitin Carboxyl-Terminal Hydrolase 6 Protein (1122-1359Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: USP6. Accession Number: P35125. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7483