Learn More
Biomatik Corporation GPX4 anti-Human, Mouse, Rat, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07902
Encompass_Preferred
The GPX4 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse, Rat. Immunogen: Recombinant Human Phospholipid hydroperoxide glutathione peroxidase, mitochondrial protein (74-197AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:500-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Phospholipid hydroperoxide glutathione peroxidase, mitochondrial, PHGPx, Glutathione peroxidase 4, GPx-4, GSHPx-4, GPX4. UniProt: P36969. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7487