Learn More
Biomatik Corporation PKHD1 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07908
Encompass_Preferred
Pkhd1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to PKHD1. The Pkhd1 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Fibrocystin Protein (520-743Aa). Applications: ELISA, WB, IF. Target Synonyms: PKHD1. Accession Number: P08F94. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7493