Learn More
Biomatik Corporation BECN1 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07912
Encompass_Preferred
The BECN1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant Human Beclin-1 protein (1-100AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB:1:500-1:5000, IHC:1:100-1:500, IF:1:50-1:500. Alternative Names: Beclin-1, Coiled-coil myosin-like BCL2-interacting protein, Protein GT197, Beclin-1-C 35 kDa, BECN1, GT197. UniProt: Q14457. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7497