Learn More
Biomatik Corporation TRADD anti-Human, Rat, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07918
Encompass_Preferred
Tradd Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to TRADD. The Tradd Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Tumor Necrosis Factor Receptor Type 1-Associated Death Domain Protein (1-312Aa). Applications: ELISA, WB, IHC, IF, IP. Target Synonyms: TRADD. Accession Number: Q15628. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7503