Learn More
Biomatik Corporation NIPBL anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07921
Encompass_Preferred
Nipbl Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to NIPBL. The Nipbl Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Nipped-B-Like Protein (2523-2697Aa). Applications: ELISA, WB, IHC. Target Synonyms: NIPBL. Accession Number: Q6KC79. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7506