Learn More
Biomatik Corporation TTC27 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07922
Encompass_Preferred
Ttc27 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to TTC27. The Ttc27 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Tetratricopeptide Repeat Protein 27 Protein (590-817Aa). Applications: ELISA, WB, IHC. Target Synonyms: TTC27. Accession Number: Q6P3X3. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7507