Learn More
Biomatik Corporation RBBP6 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07924
Encompass_Preferred
Rbbp6 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to RBBP6. The Rbbp6 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human E3 Ubiquitin-Protein Ligase Rbbp6 Protein (51-97Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: RBBP6. Accession Number: Q7Z6E9. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7509