Learn More
Biomatik Corporation SULF2 anti-Human, Mouse, Rat, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507510
Encompass_Preferred
Sulf2 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to SULF2. The Sulf2 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Extracellular Sulfatase Sulf-2 Protein (463-660Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: SULF2. Accession Number: Q8IWU5. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7510 Supplier Biomatik Corporation Supplier No. CAC07925
Sulf2 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to SULF2. The Sulf2 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Extracellular Sulfatase Sulf-2 Protein (463-660Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: SULF2. Accession Number: Q8IWU5. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.