Learn More
Biomatik Corporation DYNC2H1 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07926
Encompass_Preferred
The DYNC2H1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant Human Cytoplasmic dynein 2 heavy chain 1 protein (1928-2065AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:500-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Cytoplasmic dynein 2 heavy chain 1, Cytoplasmic dynein 2 heavy chain, Dynein cytoplasmic heavy chain 2, Dynein heavy chain 11, hDHC11, Dynein heavy chain isotype 1B, DYNC2H1, DHC1B, DHC2, DNCH2, DYH1B, KIAA1997. UniProt: Q8NCM8. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7511