Learn More
Biomatik Corporation LMX1A anti-Human, Rat, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507514
Encompass_Preferred
Lmx1A Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to LMX1A. The Lmx1A Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Lim Homeobox Transcription Factor 1-Alpha Protein (9-86Aa). Applications: ELISA, WB, IHC. Target Synonyms: LMX1A. Accession Number: Q8TE12. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7514 Supplier Biomatik Corporation Supplier No. CAC07929
Lmx1A Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to LMX1A. The Lmx1A Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Lim Homeobox Transcription Factor 1-Alpha Protein (9-86Aa). Applications: ELISA, WB, IHC. Target Synonyms: LMX1A. Accession Number: Q8TE12. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.