Learn More
Biomatik Corporation STK35 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507515
Encompass_Preferred
Stk35 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to STK35. The Stk35 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Serine; Threonine-Protein Kinase 35 Protein (1-200Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: STK35. Accession Number: Q8TDR2. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7515 Supplier Biomatik Corporation Supplier No. CAC07930
Stk35 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to STK35. The Stk35 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Serine; Threonine-Protein Kinase 35 Protein (1-200Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: STK35. Accession Number: Q8TDR2. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.