Learn More
Biomatik Corporation CPT1C anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07931
Encompass_Preferred
The CPT1C Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant Human Carnitine O-palmitoyltransferase 1, brain isoform protein (699-803AA). Applications: ELISA, WB, IHC. Recommended dilution: WB: 1:500-1:5000, IHC:1:20-1:200. Alternative Names: Carnitine O-palmitoyltransferase 1, brain isoform, CPT1-B, CPT IC, Carnitine O-palmitoyltransferase I, brain isoform, CPTI-B, Carnitine palmitoyltransferase 1C, CPT1C, CATL1. UniProt: Q8TCG5. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7516