Learn More
Biomatik Corporation GNB4 anti-Human, Mouse, Rat, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07940
Encompass_Preferred
The GNB4 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse, Rat. Immunogen: Recombinant Human Guanine nucleotide-binding protein subunit beta-4 protein (105-185AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:500-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Guanine nucleotide-binding protein subunit beta-4, Transducin beta chain 4, GNB4. UniProt: Q9HAV0. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7525