Learn More
Biomatik Corporation TRIM4 anti-Human, Rat, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507527
Encompass_Preferred
Trim4 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to TRIM4. The Trim4 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human E3 Ubiquitin-Protein Ligase Trim4 Protein (1-118Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: TRIM4. Accession Number: Q9C037. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7527 Supplier Biomatik Corporation Supplier No. CAC07942
Trim4 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to TRIM4. The Trim4 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human E3 Ubiquitin-Protein Ligase Trim4 Protein (1-118Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: TRIM4. Accession Number: Q9C037. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.