Learn More
Biomatik Corporation PIPOX anti-Human, Mouse, Rat, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07943
Encompass_Preferred
Pipox Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to PIPOX. The Pipox Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Peroxisomal Sarcosine Oxidase Protein (259-337Aa). Applications: ELISA, WB, IHC. Target Synonyms: PIPOX. Accession Number: Q9P0Z9. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7528