Learn More
Biomatik Corporation ELOVL4 anti-Human, Rat, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507531
Encompass_Preferred
The ELOVL4 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Rat. Immunogen: Recombinant Human Elongation of very long chain fatty acids protein 4 protein (270-314AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:500-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Elongation of very long chain fatty acids protein 4, 3-keto acyl-CoA synthase ELOVL4, ELOVL4. UniProt: Q9GZR5. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7531 Supplier Biomatik Corporation Supplier No. CAC07946
The ELOVL4 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Rat. Immunogen: Recombinant Human Elongation of very long chain fatty acids protein 4 protein (270-314AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:500-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Elongation of very long chain fatty acids protein 4, 3-keto acyl-CoA synthase ELOVL4, ELOVL4. UniProt: Q9GZR5. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.