Learn More
Biomatik Corporation TMCO1 anti-Human, Mouse, Rat, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07950
Encompass_Preferred
Tmco1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to TMCO1. The Tmco1 Polyclonal Antibody has been validated for the following applications: ELISA, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Calcium Load-Activated Calcium Channel Protein (43-79Aa). Applications: ELISA, IHC, IF. Target Synonyms: TMCO1. Accession Number: Q9UM00. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7535