Learn More
Biomatik Corporation CNTN6 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507536
Encompass_Preferred
The CNTN6 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant Human Contactin-6 protein (706-812AA). Applications: ELISA, WB, IHC. Recommended dilution: WB: 1:500-1:5000, IHC:1:20-1:200. Alternative Names: Contactin-6, Neural recognition molecule NB-3, hNB-3, CNTN6. UniProt: Q9UQ52. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7536 Supplier Biomatik Corporation Supplier No. CAC07951
The CNTN6 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant Human Contactin-6 protein (706-812AA). Applications: ELISA, WB, IHC. Recommended dilution: WB: 1:500-1:5000, IHC:1:20-1:200. Alternative Names: Contactin-6, Neural recognition molecule NB-3, hNB-3, CNTN6. UniProt: Q9UQ52. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.