Learn More
Biomatik Corporation VAV3 anti-Human, Mouse, Rat, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07952
Encompass_Preferred
Vav3 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to VAV3. The Vav3 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Guanine Nucleotide Exchange Factor Vav3 Protein (562-674Aa). Applications: ELISA, WB, IHC. Target Synonyms: VAV3. Accession Number: Q9UKW4. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7537