Learn More
Biomatik Corporation UBQLN1 anti-Human, Mouse, Rat, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507539
Encompass_Preferred
Ubqln1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to UBQLN1. The Ubqln1 Polyclonal Antibody has been validated for the following applications: ELISA, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Ubiquilin-1 Protein (1-300Aa). Applications: ELISA, IHC, IF. Target Synonyms: UBQLN1. Accession Number: Q9UMX0. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7539 Supplier Biomatik Corporation Supplier No. CAC07954
Ubqln1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to UBQLN1. The Ubqln1 Polyclonal Antibody has been validated for the following applications: ELISA, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Ubiquilin-1 Protein (1-300Aa). Applications: ELISA, IHC, IF. Target Synonyms: UBQLN1. Accession Number: Q9UMX0. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.