Learn More
Biomatik Corporation SLCO1B1 anti-Human, Mouse, Rat, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07959
Encompass_Preferred
Slco1B1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to SLCO1B1. The Slco1B1 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Solute Carrier Organic Anion Transporter Family Member 1B1 Protein (426-537Aa). Applications: ELISA, WB, IHC. Target Synonyms: SLCO1B1. Accession Number: Q9Y6L6. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7544