Learn More
Biomatik Corporation RUVBL2 anti-Human, Mouse, Rat, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07961
Encompass_Preferred
Ruvbl2 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to RUVBL2. The Ruvbl2 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Ruvb-Like 2 Protein (281-444Aa). Applications: ELISA, WB, IHC. Target Synonyms: RUVBL2. Accession Number: Q9Y230. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7546