Learn More
Biomatik Corporation CELF2 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507565
Encompass_Preferred
The CELF2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant Human CUGBP Elav-like family member 2 protein (291-381AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB:1:500-1:5000, IHC:1:500-1:1000, IF:1:50-1:500. Alternative Names: CUGBP Elav-like family member 2, CELF-2, Bruno-like protein 3, CUG triplet repeat RNA-binding protein 2, CUG-BP2, CUG-BP- and ETR-3-like factor 2, ELAV-type RNA-binding protein 3, ETR-3, Neuroblastoma apoptosis-related RNA-binding protein, hNAPOR, RNA-binding protein BRUNOL-3, CELF2, BRUNOL3, CUGBP2, ETR3, NAPOR. UniProt: O95319. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7565 Supplier Biomatik Corporation Supplier No. CAC07980
The CELF2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant Human CUGBP Elav-like family member 2 protein (291-381AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB:1:500-1:5000, IHC:1:500-1:1000, IF:1:50-1:500. Alternative Names: CUGBP Elav-like family member 2, CELF-2, Bruno-like protein 3, CUG triplet repeat RNA-binding protein 2, CUG-BP2, CUG-BP- and ETR-3-like factor 2, ELAV-type RNA-binding protein 3, ETR-3, Neuroblastoma apoptosis-related RNA-binding protein, hNAPOR, RNA-binding protein BRUNOL-3, CELF2, BRUNOL3, CUGBP2, ETR3, NAPOR. UniProt: O95319. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.