Learn More
Biomatik Corporation DNASE1 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07981
Encompass_Preferred
The DNASE1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant Human Deoxyribonuclease-1 protein (78-231AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-5000, IHC:1:500-1:1000. Alternative Names: Deoxyribonuclease-1, Deoxyribonuclease I, DNase I, Dornase alfa, DNASE1, DNL1, DRNI. UniProt: P24855. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7566