Learn More
Biomatik Corporation KIDINS220 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07996
Encompass_Preferred
Kidins220 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to KIDINS220. The Kidins220 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Kinase D-Interacting Substrate Of 220 Kda Protein (1077-1370Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: KIDINS220. Accession Number: Q9ULH0. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7581