Learn More
Biomatik Corporation MAP3K13 anti-Human, Mouse, Rat, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07998
Encompass_Preferred
Map3K13 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to MAP3K13. The Map3K13 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Mitogen-Activated Protein Kinase Kinase Kinase 13 Protein (506-835Aa). Applications: ELISA, WB, IHC. Target Synonyms: MAP3K13. Accession Number: O43283. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7583