Learn More
Biomatik Corporation MBP anti-Human, Mouse, Rat, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07999
Encompass_Preferred
Mbp Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to MBP. The Mbp Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Myelin Basic Protein (1-197Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: MBP. Accession Number: P02686. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7584