Learn More
Biomatik Corporation MYF6 anti-Human, Mouse, Rat, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507586
Encompass_Preferred
Myf6 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to MYF6. The Myf6 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Myogenic Factor 6 Protein (26-232Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: MYF6. Accession Number: P23409. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7586 Supplier Biomatik Corporation Supplier No. CAC08001
Myf6 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to MYF6. The Myf6 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Myogenic Factor 6 Protein (26-232Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: MYF6. Accession Number: P23409. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.