Learn More
Biomatik Corporation PER3 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507593
Encompass_Preferred
Per3 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to PER3. The Per3 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Period Circadian Protein Homolog 3 Protein (1-131Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: PER3. Accession Number: P56645. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7593 Supplier Biomatik Corporation Supplier No. CAC08008
Per3 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to PER3. The Per3 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Period Circadian Protein Homolog 3 Protein (1-131Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: PER3. Accession Number: P56645. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.