Learn More
Biomatik Corporation PRKACB anti-Human, Mouse, Rat, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507602
Encompass_Preferred
Prkacb Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to PRKACB. The Prkacb Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Camp-Dependent Protein Kinase Catalytic Subunit Beta Protein (2-53Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: PRKACB. Accession Number: P22694. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7602 Supplier Biomatik Corporation Supplier No. CAC08017
Prkacb Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to PRKACB. The Prkacb Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Camp-Dependent Protein Kinase Catalytic Subunit Beta Protein (2-53Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: PRKACB. Accession Number: P22694. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.