Learn More
Biomatik Corporation PRPS2 anti-Human, Rat, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507605
Encompass_Preferred
Prps2 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to PRPS2. The Prps2 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Ribose-Phosphate Pyrophosphokinase 2 Protein (11-106Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: PRPS2. Accession Number: P11908. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7605 Supplier Biomatik Corporation Supplier No. CAC08020
Prps2 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to PRPS2. The Prps2 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Ribose-Phosphate Pyrophosphokinase 2 Protein (11-106Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: PRPS2. Accession Number: P11908. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.