Learn More
Biomatik Corporation PTF1A anti-Human, Mouse, Rat, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507609
Encompass_Preferred
Ptf1A Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to PTF1A. The Ptf1A Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Pancreas Transcription Factor 1 Subunit Alpha Protein (224-325Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: PTF1A. Accession Number: Q7RTS3. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7609 Supplier Biomatik Corporation Supplier No. CAC08024
Ptf1A Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to PTF1A. The Ptf1A Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Pancreas Transcription Factor 1 Subunit Alpha Protein (224-325Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: PTF1A. Accession Number: Q7RTS3. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.