Learn More
Biomatik Corporation SLC16A8 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08027
Encompass_Preferred
Slc16A8 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to SLC16A8. The Slc16A8 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Monocarboxylate Transporter 3 Protein (432-504Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: SLC16A8. Accession Number: O95907. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7612