Learn More
Biomatik Corporation STAT4 anti-Human, Mouse, Rat, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507615
Encompass_Preferred
Stat4 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to STAT4. The Stat4 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Signal Transducer And Activator Of Transcription 4 Protein (569-748Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: STAT4. Accession Number: Q14765. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7615 Supplier Biomatik Corporation Supplier No. CAC08030
Stat4 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to STAT4. The Stat4 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Signal Transducer And Activator Of Transcription 4 Protein (569-748Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: STAT4. Accession Number: Q14765. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.