Learn More
Biomatik Corporation RCAN2 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08044
Encompass_Preferred
Rcan2 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to RCAN2. The Rcan2 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Calcipressin-2 Protein (88-197Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: RCAN2. Accession Number: Q14206. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7628