Learn More
Biomatik Corporation NPSR1 anti-Human, Mouse, Rat, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08055
Encompass_Preferred
Npsr1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to NPSR1. The Npsr1 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Neuropeptide S Receptor Protein (1-52Aa). Applications: ELISA, WB, IHC. Target Synonyms: NPSR1. Accession Number: Q6W5P4. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7639