Learn More
Biomatik Corporation GPIHBP1 anti-Human, Mouse, Rat, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507644
Encompass_Preferred
The GPIHBP1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse, Rat. Immunogen: Recombinant Human Glycosylphosphatidylinositol-anchored high density lipoprotein-binding protein 1 protein (21-151AA). Applications: ELISA, WB, IHC. Recommended dilution: WB: 1:500-1:5000, IHC:1:20-1:200. Alternative Names: Glycosylphosphatidylinositol-anchored high density lipoprotein-binding protein 1, GPI-HBP1, GPI-anchored HDL-binding protein 1, High density lipoprotein-binding protein 1, GPIHBP1, HBP1. UniProt: Q8IV16. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7644 Supplier Biomatik Corporation Supplier No. CAC08060
The GPIHBP1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse, Rat. Immunogen: Recombinant Human Glycosylphosphatidylinositol-anchored high density lipoprotein-binding protein 1 protein (21-151AA). Applications: ELISA, WB, IHC. Recommended dilution: WB: 1:500-1:5000, IHC:1:20-1:200. Alternative Names: Glycosylphosphatidylinositol-anchored high density lipoprotein-binding protein 1, GPI-HBP1, GPI-anchored HDL-binding protein 1, High density lipoprotein-binding protein 1, GPIHBP1, HBP1. UniProt: Q8IV16. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.