Learn More
Biomatik Corporation FUNDC1 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08062
Encompass_Preferred
The FUNDC1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant Human FUN14 domain-containing protein 1 protein (1-46AA). Applications: ELISA, WB, IHC. Recommended dilution: WB: 1:500-1:5000, IHC:1:20-1:200. Alternative Names: FUN14 domain-containing protein 1, FUNDC1. UniProt: Q8IVP5. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7646