Learn More
Biomatik Corporation ATAD1 anti-Human, Mouse, Rat, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507657
Encompass_Preferred
The ATAD1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse, Rat. Immunogen: Recombinant Human ATPase family AAA domain-containing protein 1 protein (289-350AA). Applications: ELISA, WB, IHC. Recommended dilution: WB: 1:500-1:5000, IHC:1:20-1:200. Alternative Names: ATPase family AAA domain-containing protein 1, Thorase, ATAD1, FNP001. UniProt: Q8NBU5. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7657 Supplier Biomatik Corporation Supplier No. CAC08073
The ATAD1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse, Rat. Immunogen: Recombinant Human ATPase family AAA domain-containing protein 1 protein (289-350AA). Applications: ELISA, WB, IHC. Recommended dilution: WB: 1:500-1:5000, IHC:1:20-1:200. Alternative Names: ATPase family AAA domain-containing protein 1, Thorase, ATAD1, FNP001. UniProt: Q8NBU5. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.