Learn More
Biomatik Corporation ST6GALNAC1 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08077
Encompass_Preferred
St6Galnac1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to ST6GALNAC1. The St6Galnac1 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Alpha-N-Acetylgalactosaminide Alpha-2,6-Sialyltransferase 1 Protein (36-249Aa). Applications: ELISA, WB, IHC. Target Synonyms: ST6GALNAC1. Accession Number: Q9NSC7. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7661