Learn More
Biomatik Corporation FOXQ1 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507665
Encompass_Preferred
The FOXQ1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant Human Forkhead box protein Q1 protein (10-123AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:500-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Forkhead box protein Q1, HNF-3/forkhead-like protein 1, HFH-1, Hepatocyte nuclear factor 3 forkhead homolog 1, FOXQ1, HFH1. UniProt: Q9C009. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7665 Supplier Biomatik Corporation Supplier No. CAC08081
The FOXQ1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant Human Forkhead box protein Q1 protein (10-123AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:500-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Forkhead box protein Q1, HNF-3/forkhead-like protein 1, HFH-1, Hepatocyte nuclear factor 3 forkhead homolog 1, FOXQ1, HFH1. UniProt: Q9C009. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.