Learn More
Biomatik Corporation HAO2 anti-Human, Mouse, Rat, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507667
Encompass_Preferred
The HAO2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse, Rat. Immunogen: Recombinant Human Hydroxyacid oxidase 2 protein (2-200AA). Applications: ELISA, WB, IHC. Recommended dilution: WB: 1:500-1:5000, IHC:1:20-1:200. Alternative Names: Hydroxyacid oxidase 2, HAOX2, (S)-2-hydroxy-acid oxidase, peroxisomal, Cell growth-inhibiting gene 16 protein, Long chain alpha-hydroxy acid oxidase, Long-chain L-2-hydroxy acid oxidase, HAO2, HAOX2, GIG16. UniProt: Q9NYQ3. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7667 Supplier Biomatik Corporation Supplier No. CAC08083
The HAO2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse, Rat. Immunogen: Recombinant Human Hydroxyacid oxidase 2 protein (2-200AA). Applications: ELISA, WB, IHC. Recommended dilution: WB: 1:500-1:5000, IHC:1:20-1:200. Alternative Names: Hydroxyacid oxidase 2, HAOX2, (S)-2-hydroxy-acid oxidase, peroxisomal, Cell growth-inhibiting gene 16 protein, Long chain alpha-hydroxy acid oxidase, Long-chain L-2-hydroxy acid oxidase, HAO2, HAOX2, GIG16. UniProt: Q9NYQ3. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.