Learn More
Biomatik Corporation SLC6A13 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08086
Encompass_Preferred
Slc6A13 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to SLC6A13. The Slc6A13 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Sodium- And Chloride-Dependent Gaba Transporter 2 Protein (1-40Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: SLC6A13. Accession Number: Q9NSD5. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7670