Learn More
Biomatik Corporation ETV7 anti-Human, Rat, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08092
Encompass_Preferred
The ETV7 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Rat. Immunogen: Recombinant Human Transcription factor ETV7 protein (134-218AA). Applications: ELISA, WB, IHC. Recommended dilution: WB: 1:500-1:5000, IHC:1:20-1:200. Alternative Names: Transcription factor ETV7, ETS translocation variant 7, ETS-related protein Tel2, Tel-related Ets factor, Transcription factor Tel-2, ETV7, TEL2, TELB, TREF. UniProt: Q9Y603. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7676