Learn More
Biomatik Corporation NRXN1 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08098
Encompass_Preferred
Nrxn1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to NRXN1. The Nrxn1 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Neurexin-1-Beta Protein (184-363Aa). Applications: ELISA, WB, IF. Target Synonyms: NRXN1. Accession Number: P58400. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7682