Learn More
Biomatik Corporation CCDC8 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08102
Encompass_Preferred
The CCDC8 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Coiled-coil domain-containing protein 8 protein (214-513AA). Applications: ELISA, WB, IF. Recommended dilution: WB: 1:2000-1:5000, IF:1:50-1:200. Alternative Names: Coiled-coil domain-containing protein 8, CCDC8. UniProt: Q9H0W5. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7686