Learn More
Biomatik Corporation AMACR anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08104
Encompass_Preferred
The AMACR Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Alpha-methylacyl-CoA racemase protein (13-175AA). Applications: ELISA, WB, IF. Recommended dilution: WB: 1:2000-1:5000, IF:1:50-1:200. Alternative Names: Alpha-methylacyl-CoA racemase, 2-methylacyl-CoA racemase, AMACR. UniProt: Q9UHK6. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7688