Learn More
Biomatik Corporation C5AR1 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08105
Encompass_Preferred
The C5AR1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human C5a anaphylatoxin chemotactic receptor 1 protein (1-36AA). Applications: ELISA, WB, IF. Recommended dilution: WB: 1:1000-1:5000, IF:1:50-1:200. Alternative Names: C5a anaphylatoxin chemotactic receptor 1, C5a anaphylatoxin chemotactic receptor, C5a-R, C5aR, CD88, C5AR1, C5AR, C5R1. UniProt: P21730. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7689